

Diisopropyl ether ≥98%, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:108-20-3
:
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880
:102.18 g/mol
Boiling Pt:68,5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0,726 g/cm³ (20 °C)
Flash Pt:−28 °C
EINECS:203-560-6
UN:1159
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Guarantee Analysis:.
- Assay(GC):Min. 98.00 %
- Weight Per ml (at 20°C):0.722 --> 0.724 g
- Refractive Index (at 20°C):1.368 --> 1.369
- Maximum Limits Of Impurities:Jun-03
- Peroxide(As C6H14O2):Max. 0.05 %
- Residue After Evaporation:Max. 0.01 %
- Water ( By KF):Max. 0.10 %
Specification Test Results
Related Information
Frequently Bought Together