Order Entry
India
Orders LinkContactUsLinkComponent
VWR® Microtips
Doublestep micro tip, lower portion only, 3 mm
  VWRU33996-265
VWR® Microtips
  VWRU33996-265
VWR® Microtips
VWR® Microtips
Doublestep micro tip, lower portion only, 3 mm
Rs.80,012.00
1 * 1 items
Microtips made of titanium for use with Sonifier® cell disrupters.
The double stepped microtip assembly consists of a coupler and a stepped tip and attaches directly without need for a tapped horn. It has lower amplitude than the tapered tip, but is capable of reaching into small diameter vessels of greater depth and can process solutions ≤1 µl.
  • Description:
    Doublestep micro tip, lower portion only, 3 mm