



Mercury(II) chloride ≥99.5% AR ACS, Analytical reagent (AR), Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:7487-94-7
:
Formula:HgCl₂
MDL Number:MFCD00011041
:271.5 g/mol
Melting Pt:273 °C
Density:5,44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-299-8
UN:1624
ADR:6.1,II
:GHS05
:GHS06
:GHS08
:GHS09
Specification Test Results
- Solution In Ethyl Ether:Passes test
- Guarantee Analysis:.
- Assay (HgCl2):Min. 99.50 %
- Identity:Passes test
- Maximum Limits Of Impurities:.
- Residue After Reduction:Max. 0.02 %
- Mercurous Chloride (Cl):Max. 0.05 %
- Iron (Fe):<=0.0005%
- Nitrate (NO3):<=0.0005%
- Cadmium (Cd):Max. 0.0005 %
- Calcium (Ca):Max. 0.002 %
- Copper(Cu):Max. 0.0005 %
- Lead ( Pb):Max. 0.001 %
- Magnesium (Mg):Max. 0.002 %
- Potassium (K):Max. 0.005 %
- Sodium (Na):Max. 0.005 %
- Zinc (Zn):Max. 0.001 %
Specification Test Results
Related Information
Frequently Bought Together