


Mercury(II) nitrate 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
31530.295
:
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Titer (20°C; real value 0.2 % accuracy):0.0499 - 0.0501 mol/l
- NIST traceable:Confirmed
:
:324.60 g/mol
:1,020 g/cm³ (20 °C)
:233-152-3
:3287
:6.1,II
:GHS06
:GHS08
:GHS09
:10045-94-0
:HgN₂O₆
:MFCD00011038