Hydrochloric acid 0.03 mol/l (0.03 N) (R2) Reag. Ph. Eur. 1043505
Catalog # 87843.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Density:1,004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
:36.46 g/mol
UN:1789
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324