Order Entry
India
ContactUsLinkComponent
Triethylamine ≥99.80% for HPLC, Rankem®
Flammable
Irritant
Corrosive
Triethylamine ≥99.80% for HPLC, Rankem®
Supplier:  RANKEM
Certificates
Flammable
Irritant
Corrosive
Triethylamine ≥99.80% for HPLC, Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.80%
  • Weight Per ml (at 20 °C):
    0.726 --> 0.728 g
  • Maximum Limits Of Impurities:
    .
  • Water (By KF):
    Max. 0.08%
  • Non volatile matter:
    Max. 0.003%
  • UV Transmittance:
    .
  • Wavelength:
    .
  • 330 nm:
    Min. 60%
  • 360 nm:
    Min. 80%
  • 380 nm:
    Min. 90%
  • 390 nm:
    Min. 95%
  • 400 nm:
    Min. 98%

Specification Test Results

Related Information

Frequently Bought Together