


N,N-Dimethylformamide ≥99.80% for HPLC, Rankem®
Catalog # RANKD0133
Supplier: RANKEM
CAS Number:
Specifications
- Pack type:Amber colored glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Guarantee Analysis:.
- Assay(GC):Min. 99.80%
- Maximum Limits Of Impurities:.
- Acidity(As HCOOH):Max. 0.003%
- Water ( By KF):Max. 0.05%
- Non volatile matter:Max. 0.0005%
- UV Transmittance:.
- Wavelength:.
- 275 nm:Min. 50%
- 290 nm:Min. 80%
- 300 nm:Min. 90%
- 330 nm:Min. 98%
Specification Test Results
About this item
:
:73.09 g/mol
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–60,5 °C
Density:0,949 g/cm³ (20 °C)
Flash Pt:57,5 °C (DIN 51755) (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:GHS02
:GHS07
:GHS08
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284