Order Entry
India
ContactUsLinkComponent
1-Propanol 99.8 % + for HPLC, Rankem®
Flammable
Corrosive
Irritant
1-Propanol 99.8 % + for HPLC, Rankem®
Catalog # RANKP0646
Supplier:  RANKEM
CAS Number:  
Flammable
Corrosive
Irritant
1-Propanol 99.8 % + for HPLC, Rankem®
Catalog # RANKP0646
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pack type:
    Amber colored glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Colour (APHA):
    ≤10
  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.80%
  • Weight Per ml (at 20 °C):
    0.803 --> 0.806 g
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    Max. 0.0005%
  • Titrable Acid:
    Max. 0.0002 meq/g
  • Titrable Base:
    Max. 0.0002 meq/g
  • Water ( By KF):
    Max. 0.05%
  • UV Transmittance:
    .
  • Wavelength:
    .
  • 230 nm:
    Min. 50%
  • 240 nm:
    Min. 80%
  • From 275 nm:
    Min. 98%

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:12,08027
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–127 °C
Density:0,8053 g/cm³ (20 °C. DIN 51757)
Flash Pt:15 °C
EINECS:200-746-9
UN:1274
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:71-23-8
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941