



n-Hexane 99%+ for HPLC, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94,3 °C
Density:0,659 g/cm³ (20 °C)
Flash Pt:–22 °C
EINECS:203-777-6
UN:1208
ADR:3,II
REACH:01-2119480412-44
:GHS02
:GHS08
:GHS06
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Color (APHA):≤ 10
- Guarantee Analysis:.
- Assay(GC):Min. 99.00 %
- Weight Per ml (at 20°C):0.659 -- 0.660 g
- Identity (IR):Conforms
- Maximum Limits Of Impurities:.
- Residue After Evaporation:Max. 0.0005 %
- Titrable Acid:Max. 0.0002 meq/g
- Titrable Base:Max. 0.0002 meq/g
- Water ( By KF):Max. 0.02 %
- UV Transmittance:.
- Wavelength:.
- 210 nm:Min. 60 %
- 220 nm:Min. 80 %
- 230 nm:Min. 90 %
- From 245 nm:Min. 98 %
Specification Test Results
Related Information
Frequently Bought Together