Glycerine ≥99.0%, liquid
Catalog # RANKG0060B
Supplier: RANKEM
CAS Number:
Specifications
- Pk:25 L
- Seq:5327
Specifications
Specification Test Results
- Grade standard:LR grade
Specification Test Results
About this item
:
:92.09 g/mol
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722