Order Entry
India
ContactUsLinkComponent
BeneSphera™ H31 Plus Diluent, 3 Part Differential
BeneSphera™ H31 Plus Diluent, 3 Part Differential
  BENP15001483
 :  
BeneSphera™ H31 Plus Diluent, 3 Part Differential
  BENP15001483
 :  
  • Pk:
    20 L
  • Description:
    BeneSphera™ H31 plus diluent, 3 part differential
For diluting blood samples in fully automated 3 part analyser.
  • Stable
  • Unique packaging
  • Synergic with fully automated heamtology analyser
 : Avoid the contact with eyes and skin.