Order Entry
India
ContactUsLinkComponent
BeneSphera™ H33S ProbeCleaner, 3 Part Differential
BeneSphera™ H33S ProbeCleaner, 3 Part Differential
Catalog # BENP15001301
Supplier:  BENESPHERA
CAS Number:  
BeneSphera™ H33S ProbeCleaner, 3 Part Differential
Catalog # BENP15001301
Supplier:  BENESPHERA
CAS Number:  

Specifications

  • Pk:
    20 mL
  • Description:
    BeneSphera™ H33S Probe cleaner, 3 part differential
  • Seq:
    03

Specifications

About this item

For cleaning of probe of fully automatated heamatology analyser.

  • Best fit for 3 part hematology analyser
  • Storage at RT
  • Long shelf life
Caution: Avoid the contact with eyes and skin.