Order Entry
India
ContactUsLinkComponent
BeneSphera™ H-33S Lyse, 3 Part Differential
BeneSphera™ H-33S Lyse, 3 Part Differential
Catalog # BENP15001300
Supplier:  BENESPHERA
CAS Number:  
BeneSphera™ H-33S Lyse, 3 Part Differential
Catalog # BENP15001300
Supplier:  BENESPHERA
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    BeneSphera™ H-33S lyse, 3 part differential
  • Seq:
    04

Specifications

About this item

For determination of HB, counting and sizing of lucocytes in hematology analyser.

  • Best fit for 3 part hematology analyser
  • Storage at RT
  • Long shelf life
Caution: Avoid the contact with eyes and skin.