BeneSphera™ H-33S Diluent, 3 Part Differential
Catalog # BENP15001299
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:20 L
- Description:BeneSphera™ H-33S diluent, 3 part differential
- Seq:05
Specifications
About this item
For diluting blood samples in fully automated 3 part analyser.
- Stable
- Unique packaging
- Synergic with fully automated heamtology analyser
Caution: Avoid the contact with eyes and skin.