Order Entry
India
ContactUsLinkComponent
BeneSphera™ H33S LYse 3 Part Differential-RFID
BeneSphera™ H33S LYse 3 Part Differential-RFID
Catalog # BENP15001486
Supplier:  BENESPHERA
CAS Number:  
BeneSphera™ H33S LYse 3 Part Differential-RFID
Catalog # BENP15001486
Supplier:  BENESPHERA
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    BeneSphera™ H33S LYse 3-part differential-RFID

Specifications

About this item

For determination of HB, counting and sizing of lucocytes in hematology analyser.

  • Best fit for 3 part hematology analyser
  • Storage at RT
  • Long shelf life
Caution: Avoid the contact with eyes and skin.