BeneSphera™ Rinse, 3 Part Differential
Catalog # BENP15001082
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:20 L
- Description:BeneSphera™ rinse, 3 part differential
- Seq:03
Specifications
About this item
Hematology rinse solution PH 7,00 for 3 part hematology analyser.
- Best fit for 3 part hematology analyser
- Storage at RT
- Long shelf life
Caution: Avoid the contact with eyes and skin.