BeneSphera™ CF Lyse, 3 Part Differential
BENP15001081
:
- Pk:500 mL
- Description:BeneSphera™ CF lyse, 3 part differential
- Seq:01
For diluting blood samples in fully automated 3 part analyser.
- Best fit for 3 part hematology analyser
- Storage at RT
- Long shelf life
: Avoid the contact with eyes and skin.