Order Entry
India
ContactUsLinkComponent
VWR® Screw Top Vial, 9 mm
VWR® Screw Top Vial, 9 mm
Catalog # 548-2599
Supplier:  VWR Collection
VWR® Screw Top Vial, 9 mm
Catalog # 548-2599
Supplier:  VWR Collection

Specifications

  • Type:
    Micro vial
  • Capacity:
    0,3 ml (300 µl)
  • Material:
    PP
  • Pk:
    100

Specifications

About this item