Benesphera™ EZ cleaner, 3 part differential analyser
Catalog # BENP15001083
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:100 mL
- Description:Benesphera™ EZ cleaner, 3 part differential analyser
- Seq:05
Specifications
About this item
For Bath before shut down in hematology analyzer
- Best Fit for 3 Part Hematology analyzer
- Storage at RT
- Long Shelf Life
Avoid the contact with eyes & Skin