BeneSphera™ ESR Pipettes
BENP16008369
- Description:BeneSphera™ ESR Pipettes
- Environmentally Preferable:
- Pk:1
BeneSphera ESR Tubes are single use devices intended to be used as whole blood sample tubes for the quantitative determination of Erythrocyte Sedimentation Rate (ESR).
- Certified Calibration of Accuracy and Precise Graduations
- Clear, Bold, Bi-directional Graduations
- Filter Plug - Prevent aerosol or liquid contamination of the pipetting device while further reducing sample-to-sample contamination risks
- Negative Graduations - Increase pipette capacity, and accommodates additional volumes
An Erythrocyte Sedimentation Rate (ESR) is a test for finding out presence of inflammation or infections in the body.