Order Entry
India
ContactUsLinkComponent
Biuret reagent solution Reag. Ph. Eur.
Irritant
Biuret reagent solution Reag. Ph. Eur.
Catalog # 87787.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Biuret reagent solution Reag. Ph. Eur.
Catalog # 87787.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3266
ADR:8,II
 :GHS07
 :108-19-0