Order Entry
India
ContactUsLinkComponent
 
Tetrahydrofuran, ultra-low water content ≥99.8%, BakerDRY™, J.T.Baker®
Catalog # 9447-12
Supplier:  Avantor
CAS Number:  
undefined
Tetrahydrofuran, ultra-low water content ≥99.8%, BakerDRY™, J.T.Baker®
Catalog # 9447-12
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth, septum-sealed
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • ACS - Assay (by GC, corrected for water):
    ≥99.8 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Peroxide (as H₂O₂) (at time of manufacture):
    ≤0.015 %
  • ACS - Residue after Evaporation:
    ≤0.03 %
  • ACS - Water (by KF, coulometric):
    ≤10.0 ppm
  • Stabilizer (BHT):
    100 - 300 ppm

Specification Test Results

About this item

Synonyms: Butylene oxide, THF, Diethylene oxide
 :
 :72.11 g/mol
EINECS:203-726-8
UN:2056
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356
Boiling Pt:66
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
ADR:3 II
Merck Index:12,09351