Toluidine blue O, high purity
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:92-31-9
:
Formula:C₁₅H₁₆ClN₃S
MDL Number:MFCD00011934
:305.83 g/mol
Merck Index:12,09658
Storage Temperature:Ambient
EINECS:202-146-2
Specification Test Results
- Dye Content:>50.0%
- Loss on Drying:≤8.0%
- Color Index:52040
Specification Test Results
Frequently Bought Together