pH Test Strips, VWR Chemicals BDH®
VWRUBDH83930.601
- pH gradation:0.5
- pH range:0 - 6.0
- Pk:1
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use.
- Designed for safe testing of dangerous, poisonous, or aggressive liquids
- Long handle protects user from contact with sample
- Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions
Dimensions: (L×W) 85×6 mm.
: Strips are packaged 100 to a box. Included colour chart has four different color blocks for each pH value