Order Entry
India
ContactUsLinkComponent
Hematocrit Centrifuge, iFuge HCT NXT
Hematocrit Centrifuge, iFuge HCT NXT
  NEUAIFUGEHCT-NXT
Hematocrit Centrifuge, iFuge HCT NXT
  NEUAIFUGEHCT-NXT
  • Max. speed:
    500 - 12000 min⁻ ¹
  • Description:
    Hematocrit centrifuge
  • Rotor type:
    HCT rotor with screw on lid
  • Max. RCF:
    14118 g
  • Dimensions:
    330×265×161 mm
  • Display:
    Digital
  • Max. speed - swing-out rotor:
    12000 min⁻ ¹
  • Motor:
    BLDC
  • Timer:
    Upto 12 mins
  • Weight:
    4,5 kg
  • Pk:
    1
  • Rotors included:
    Yes
Compact hematocrit centrifuge ideal for determining erythocyte fractions in blood using its included 24 place capillary rotor. Comes with a long-lasting brushless DC motor that is designed to offer the best-in-class long term consistent performance.
  • Microprocessor controlled for efficient performance even under high load
  • Two preset feature to set favorite mode where speed and time can be changed as per requirement
  • HCT rotor with screw-on lid for better contamination protection
  • Compact design saves valuable bench space