Order Entry
India
ContactUsLinkComponent
J.T.Baker® BAKER BIO-ANALYZED®, Buffer A: 20 mm NaOH in 10% Acetonitrile
J.T.Baker® BAKER BIO-ANALYZED®, Buffer A: 20 mm NaOH in 10% Acetonitrile
Catalog # 9122-51
Supplier:  Avantor
CAS Number:  
undefined
J.T.Baker® BAKER BIO-ANALYZED®, Buffer A: 20 mm NaOH in 10% Acetonitrile
Catalog # 9122-51
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    20 mM NaOH
  • Pk:
    200 L
  • Description:
    Buffer A
  • Size:
    200 L

Specifications

About this item

  • Sodium Hydroxide (by titration) (mM): 18 to 22

Manufactured with water meeting the following requirements:

Total organic compounds (500 ppb max.)
Conductivity, (2,1 uS/cm max.)
Microbes (50 CFU max.)

Caution: For laboratory, research or manufacturing use.