Order Entry
India
ContactUsLinkComponent
BeneSphera™ H31 Plus Hematology Analyser
BeneSphera™ H31 Plus Hematology Analyser
Catalog # BENP20002001
Supplier:  BENESPHERA
undefined
BeneSphera™ H31 Plus Hematology Analyser
Catalog # BENP20002001
Supplier:  BENESPHERA
Specifications
  • Description:
    3 Part Differential Heamatology Analyzer H31 Plus
  • Pk:
    1 KIT
Specifications
About this item
Quantitative hematology analysis and 3 part differential counting for in vitro use in your laboratory. The advanced BeneSphera H31 Plus Hematology Analyzer supports identification of normal patients with system generated parameters and flags patient results that require your attention
  • CBC + DIFF , 22 paramters, 3 Histograms
  • Throughput upto 40 samples / hour
  • 10.4 inch TFT touch screen
  • Only three reagents needed
  • Detailed flag information
  • Large storage capacity upto 100,000 samples
  • Sample volume is only 9ul which is ideal for pediatric samples
Minimum Size, Maximum Capacity