Order Entry
India
ContactUsLinkComponent
BeneSphera™ H33s Hematology Analyser
  BENP20002002
undefined
BeneSphera™ H33s Hematology Analyser
  BENP20002002
  • Description:
    3 Part Differential Heamatology Analyzer H33s-RFID-(D)
  • Pk:
    1 KIT
The BeneSphera™ H33s is a compact, powerful, full featured and affordable system. It provides a detailed flag information never before seen on a 3-part analyzer. The system provides information
useful for diagnosis including WBC flag, RBC flag and PLT flag.
  • COMPACT DESIGN
  • COST EFFICIENT
  • Higher throughput at 70 tests per hour
  • MICRO SAMPLE VOLUME
  • Micro sample volume at 9.0uL for whole blood mode with capillary whole blood samples supported, perfect for pediatric samples
  • BETTER CONNECTIVITY
Next generation hematology analyzer