Order Entry
India
ContactUsLinkComponent
HPLC Analyser, GH 900 Plus
HPLC Analyser, GH 900 Plus
  LIFO120210
undefined
HPLC Analyser, GH 900 Plus
  LIFO120210
  • Description:
    GH 900 Plus HPLC analyser
  • Pk:
    1
  • Fully automated - to minimise operation steps
  • Precise and reliable - to serve you consistently (CV ≤1,5%)
  • Dual wavelength detection - to avoid interference (415 nm + 500 nm detector)
  • Degasser - For better result accuracy
  • Compact size - To minimise space requirements
HPLC technology - Gold standard methodology
- NGSP and IFCC certified
- HbA1c results within 130 seconds
- 5 samples loading facility, suitable for medium to small labs