- Description:BC 5150 Hematology Analyser
- Environmentally Preferable:
- Pk:1
A compact and user friendly, high performance 5 part hematology system. The BC 5150 Auto Hematology Analyzer is a light and compact system, specially tailored to assist diagnostic labs who need full CBC+5 part results, with a relatively low daily sample volume. Responding to increased demands
for high quality read-outs and accurate diagnostic decisions, the BC 5150 provides a reliable complete 5-part differential menu, 25 reportable parameters and 12 research parameters, 3 histograms and 3 scattergrams.
- Outstanding accuracy for a reliable diagnosis
- Fast Throughput with increased walk away
- Compact Design
- Convenience and Efficiency
- High performance technology automation
- Bi Directional LIS connectivity
The measurement methods used in this analyzer are:
• the Electrical Impedance method for determining the RBC and PLT data;
• the colorimetric method for determining the HGB;
• flow cytometry by laser for determining the WBC data.
Tri angle scatter + focussed flow + chemical dye, creates the possibility for a better 5-part WBC differentiation.