Order Entry
India
ContactUsLinkComponent
BeneSphera™ T4 MW ELISA - Novo
BeneSphera™ T4 MW ELISA - Novo
  BENP15001539
 :  
undefined
BeneSphera™ T4 MW ELISA - Novo
  BENP15001539
 :  

 

  • Pk:
    96 Tests

 

 

BeneSphera™ T4 Novo Elisa kit is for quantitative determination of T4 (thyroxine) hormone from human serum and plasma.

  • Assay time: 1 hour 30 min
  • Expected range: male: 4,4 - 10,8 µg/dl, female: 4,8 - 11,6 µg/dl
  • Sensitivity: 0,4 µg/dl
  • Specificity: No interference found, a total of 131 samples found to have high degree of correlation with T4 calibrators provided

Enzyme Immunoassay for the quantitative determination of T4 Hormone concentration in human serum/plasma - for invitro diagnostics use only.