VWR® Gamma Irradiated Triple Wrapped Polyester 70% IPA Cleanroom Wipes
Ready-to-use 100% polyester wipes in sterile packaging, pre-saturated with 70% IPA and 30% WFI.
- Ready-to-use pull-up wipes in sterile packaging can be used directly on surfaces
- Heat sealed edges with double knit structure
- Manufactured in a grade A cleanroom environment
- Triple bagged and gamma irradiated to preserve product integrity
- Packed wipes maintain sterility for up to 17 weeks after opening
Sterilised by gamma irradiation at not less than 25 kGy, these 100% polyester cleanroom wipes are pre-saturated and ready-to-use, with low particulate emission. The superior quality, non-woven, hydroentangled material offers high absorbency for liquids on any surface, making them ideal for cleaning and disinfecting critical surfaces.
: Validated per the Association for the Advancement of Medical Instrumentation (AAMI) 11137 guidelines to a 10⁻⁶ Sterility Assurance Level (SAL). ISO, GMP, and CE certified.
: Store wipes at room temperature. Certificate of analysis provided with every batch. Sticker label with product description on box as well as each pack.
: All products are cleanroom packed and triple bagged. Triple wrapped gamma irradiated ten packs are supplied in one case.
Frequently Bought Together