Order Entry
India
ContactUsLinkComponent
 
BeneSphera™ Syphilis Rapid Card Test
  BENP15001448
 :  
undefined
BeneSphera™ Syphilis Rapid Card Test
  BENP15001448
 :  

 

  • Pk:
    50 Tests
  • Description:
    BeneSphera™ Syphilis rapid card test
  • Seq:
    02

 

 

BeneSphera™ Syphilis rapid card test for the detection of antibodies to Syphilis from human serum, plasma, and whole blood.

  • No assay buffer required
  • User friendly
  • Can detect antibody from serum/plasma
  • Sensitivity: 100%
  • Specificity: 99,8%
  • Test time: 10 to 15 min

BeneSphera™ Syphilis rapid card test is a solid phase immunochromatographic assay for the rapid, qualitative, and differential detection of antibodies to Treponema pallidum in human serum or plasma. This test is intended for professional use and provides only a preliminary test result.