BeneSphera™ Malaria Pv/Pf 2.0 Rapid Card Test
BENP15001479
:
- Pk:1 KIT
- Description:BeneSphera™ Malaria Pv/Pf 2,0 rapid card test
- Tests per kit:25 tests
BeneSphera™ Malaria Pv/Pf 2.0 rapid card test (Lateral Flow) for the detection of Malaria antigen [Pv (pLDH)/ Pf (HRP2)].
- Detects and differentiates P.Falciparum and P. Vivax
- Low requirement of sample volume (4 μl)
- Detection up to 50 p/μl (WHO requirement is 200 p/μl)
- Better background clearance and line intensity
- Sensitivity: 99,1%
- Specificity: 99,6%
BeneSphera™ Malaria Pv/Pf 2.0 antigen test is a quick immunochromatographic, qualitative, sensitive and specific test for the distinct diagnosis of Pv specific LDH and Pf specific HRP2, allowing it to detect Pv and Pf distinctly.
This test is only a preliminary screening test for malaria differential diagnosis and for in vitro diagnostics use.
: National Institute of Malaria Reaserch (ICMR, Govt. of India).