BeneSphera™ Hepatitis B Rapid Card Test
Catalog # BENP15001296
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:50 Tests
- Description:BeneSphera™ Hepatitis B rapid card test
- Environmentally Preferable:
Specifications
About this item
BeneSphera™ Hepatitis B rapid card test is designed for the detection Hepatitis B surface antigen from human serum and plasma.
- Detects all major 10 serotypes of HBsAg
- Detection limit: 0,40 IU/ml HBsAg
- Better background clearance and line intensity
- No cross reactivity with Hepatitis A and Hepatitis C 0,5 ng/ml
- Sensitivity: ≥99%
- Specificity: ≥98%
BeneSphera™ Hepatitis-B Rapid Card Test is an in vitro, qualitative, one-step immunochromatographic test for the detection of Hepatitis B surface antigen in human serum or plasma.
BeneSphera™ Hepatitis-B detects all 10 major serotypes of HBV. Cross reactivity with Hepatitis A and Hepatitis C was not observed up to 0.5 ng/ml.
Certifications: National Institute of Biologicals (MoHFW, Govt. of India).