Order Entry
India
ContactUsLinkComponent
 
BeneSphera™ T3 ELISA Kit
Catalog # BENP15000964
Supplier:  BENESPHERA
CAS Number:  
undefined
BeneSphera™ T3 ELISA Kit
Catalog # BENP15000964
Supplier:  BENESPHERA
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Description:
    BeneSphera™ T3 ELISA kit

Specifications

About this item

BeneSphera™ T3 ELISA kit for the quantitative determination of T3 (Triiodothyronine) Hormone from human serum and plasma.

  • Assay time: 1 hour 30 min
  • Expected range: 0,52 to 1,85 ng/ml
  • Sensitivity: 0,04 ng/ml
  • Specificity: no interference found, total of 162 samples found to have high degree of correlation with T3 calibrators provided
  • Precision: very low coefficient of variation found in intra-assay and inter-assay reproducibility studies

Enzyme Immunoasaay for the quantitative determination of T3 Harmone concetraion in human serum/plasma. For invitro diagnostics use only.