BeneSphera™ Pro HIV 1/2 3.1 Rapid Card Test
Catalog # BENP15001480
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:50 Tests
- Description:BeneSphera™ Pro HIV 1/2 3,1 rapid card test
- Environmentally Preferable:
Specifications
About this item
BeneSphera™ Pro HIV 1/2 3.1 rapid card test is designed for the detection of antibodies against HIV 1 and HIV 2 from human whole blood, serum, and plasma.
- Detects and differentiats HIV 1 and 2
- Qualitative detection of antibody against HIV 1 (Including Group O)
- Better background clearance and line intensity
- Sensitive to detect low antibody titre
- Sensitivity: 100%
- Specificity: 99,92%
BeneSphera™ Pro HIV 1/2 Antibody 3.1 Rapid Card test is a rapid and visual immunochromatographic assay, intended for the qualitative detection of antibodies against HIV 1 (including group O) and/or HIV 2 from human whole blood, serum or plasma.
Certifications: National Institute of Biologicals (MoHFW, Govt. of India).