Order Entry
India
ContactUsLinkComponent
 
BeneSphera™ Prolactin ELISA Kit
Catalog # BENP15000978
Supplier:  BENESPHERA
CAS Number:  
undefined
BeneSphera™ Prolactin ELISA Kit
Catalog # BENP15000978
Supplier:  BENESPHERA
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Description:
    BeneSphera™ Prolactin ELISA kit

Specifications

About this item

BeneSphera™ Prolactin ELISA Kit for the quantitative determination of Prolactin Hormone from human serum and plasma.

  • Quantitative determination by sandwich ELISA
  • Assay time: 1 h 30 min
  • Sensitivity: 0,04 ng which is equivalent to a sample containing 0,8 ng/ml Prolactin concentration
  • Specificity: no cross reactivity up to 1000 ng/ml of LH, FSH, TSH, GH and CG

BeneSphera™ Prolactin was studied for intra assay and Interassay reproducibility by analysis on pooled control sera. Intra assay reproducibility gave CV of < 4.5% and for interassay was found to be < 3.5%.

Caution: For invitro diagnostics use only.