Order Entry
India
ContactUsLinkComponent
 
BeneSphera™ FSH ELISA Kit
Catalog # BENP15000974
Supplier:  BENESPHERA
CAS Number:  
undefined
BeneSphera™ FSH ELISA Kit
Catalog # BENP15000974
Supplier:  BENESPHERA
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Description:
    BeneSphera™ FSH ELISA kit

Specifications

About this item

BeneSphera™ FSH ELISA Kit for the quantitative determination of FSH (Follicle-stimulating hormone) Hormone from human serum and plasma.

  • Quantitative determination by sandwich ELISA
  • Assay time: 1 h 30 min
  • Sensitivity: 0,02 mIU/ml which is equivalent to a sample containing 0,8 mIU/ml FSH concentration
  • Specificity: no cross reactivity

BeneSphera™ FSH was studied for intra assay and Interassay reproducibility by analysis on pooled control sera. Intra assay reproducibility gave CV of < 4.5% and for interassay was found to be < 6.9%.

Caution: For invitro diagnostics use only.