BeneSphera™ FT3 ELISA Kit
Catalog # BENP15000968
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:96 Tests
- Description:BeneSphera™ FT3 ELISA kit
Specifications
About this item
BeneSphera™ FT3 ELISA Kit for the quantitative determination of FT3 (Free Triiodothyronine) Hormone from human serum and plasma.
- Assay time: 1 h 30 min
- Expected range: Adult - 1,4 to 4,2 pg/ml, Pregnancy - 1,8 to 4,2 pg/ml
- Sensitivity: 0,05 pg/dl
- Specificity: no interference found, total of 85 samples found to have high degree of correlation with free T3 calibrators provided
- Precision: very low coefficient of variation found in intra-assay and inter-assay reproducibility studies
Enzyme Immunoassay for the quantitative determination of Free T3 Hormone concentration in human serum/plasma - for invitro diagnostics use only.
Caution: For invitro diagnostics use only.