BeneSphera™ TSH ELISA Kit
BENP15000972
:
- Pk:96 Tests
- Description:BeneSphera™ TSH ELISA kit
BeneSphera™ TSH ELISA Kit for the quantitative determination of TSH (Thyroid Stimulating Hormone ) Hormone from human serum and plasma.
- Assay time: 1 h 30 min
- Expected range: 0,39 to 6,16 µIU/ml
- Sensitivity: 0,078 µIU/ml
- Specificity: no interference found, total of 170 samples found to have high degree of correlation with TSH calibrators provided
- Precision: very low coefficient of variation found in intra-assay and inter-assay reproducibility studies
Enzyme Immunoassay for the quantitative determination of TSH Hormone concentration in human serum/plasma.
: For invitro diagnostics use only.