BeneSphera™ T4 ELISA Kit
Catalog # BENP15000967
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:96 Tests
- Description:BeneSphera™ T4 ELISA kit
Specifications
About this item
BeneSphera™ T4 ELISA Kit for the quantitative determination of T4 (thyroxine) Hormone from human serum and plasma.
- Assay time: 1 h 30 min
- Expected range: male- 4,4 to 10,8 µg/dl, female- 4,8 to 11,6 µg/dl
- Sensitivity: 0,4 µg/dl
- Specificity: no interference found, total of 131 samples found to have high degree of correlation with T4 calibrators provided
Enzyme Immunoassay for the quantitative determination of T4 Hormone concentration in human serum/plasma.
Caution: For invitro diagnostics use only.