Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,N-Dimethylacetamide
N,N-Dimethylacetamide for gas chromatography
N,N-Dimethylacetamide for gas chromatography
RANKD0772
:
RANKEM
:
N,N-Dimethylacetamide for gas chromatography
RANKD0772
:
RANKEM
:
Pk:
1 L
Seq:
5037
:
:
87.12 g/mol
:
127-19-5
:
CH₃CON(CH₃)₂
:
MFCD00008686