VWR® Media Bottles, PETG and PET
Supplier: VWR Collection
Certificates
About this item
VWR® media bottles provides the complete solution for storage of media, biological buffers, and various reagents. These bottles are leakproof, shatter-proof and freezer safe making them ideal for storage and transportation.
- Manufactured under class 8 cleanroom
- Made from USP class VI raw material with low leachable extractables
- Heat-shrink packaging and temper evident packaging seal
- Suitable for frozen storage down up to –70 °C
- Gamma sterile product
- Non pyrogenic and non cytotoxic
- Graduated for quick user visibility
VWR® Media bottles are available in PETG and PET, and are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to +121 °C. Eliminate washing, depyrogenation, and autoclaving steps with these sterile and non-pyrogenic bottles. Bottles are arranged in shrink-wrapped trays and polybags to prevent contamination.
Frequently Bought Together