J.T.Baker®, Microplate Lid
Catalog # 94001-0230
Supplier: Avantor
Specifications
- Sterility:Non sterile
- Colour:Clear
- Description:PS lid for J.T.Baker® 384 and 96 well microplate
- Pk:100
Specifications
About this item
Polystyrene lid suitable for use with J.T.Baker® 384 and 96 SW.
- Material: Polystyrene
- Dimensions (outside): 129,2×86,0×9,0 mm