Sodium acetate trihydrate ≥99% F.C.C. , U.S.P
Catalog # MACR7356-02
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:136.08 g/mol
Merck Index:13,08642
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
CAS Number:6131-90-4
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557