L(+)-Leucine USP, Multi-Compendial, Macron Fine Chemicals™
:61-90-5
:
:C₆H₁₃NO₂
:MFCD00002617
:131.17 g/mol
:14,05451
:300 °C
:1,165 g/cm³ (20 °C)
:Ambient
:200-522-0
- USP - Assay (C₆H₁₃NO₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):14.9 - 17.3 Degree
- USP - pH:5.5 - 7.0
- USP - Loss on Drying at 105°C:≤ 0.2 %
- USP - Residue on Ignition:≤ 0.4 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds - Individual Impurities:≤ 0.5 %
- USP - Related Compounds - Total Impurities:≤ 2.0 %
- EP/BP - Assay (C₆H₁₃NO₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):14.5 - 16.5 Degree
- EP/BP-Ninhydrin-Positive Substances-Impurity A:≤ 0.8 %
- EP/BP-Ninhydrin-Positive Substances-Each:≤ 0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤ 1.0 %
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP/BP - Ammonium (NH₄):≤ 0.02 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying at 105°C:≤ 0.5 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- JP - Assay (C₆H₁₃NO₂) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):14.5 - 16.0 Degree
- JP - pH:5.5 - 6.5
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.021 %
- JP - Sulfate (SO₄):≤ 0.028 %
- JP - Ammonium (NH₄):≤ 0.02 %
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Arsenic (As):≤ 2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤ 0.30 %
- JP - Residue after Ignition:≤ 0.10 %
Frequently Bought Together