Rs.0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
India
ContactUsLinkComponent
REZI®
REZI® 28 ash residue remover, J.T.Baker®
REZI® 28 ash residue remover, J.T.Baker®
REZI® 28 ash residue remover, J.T.Baker®
Catalog # 6426-05
Supplier:
Avantor
CAS Number:
Expand image
REZI® 28 ash residue remover, J.T.Baker®
Catalog # 6426-05
Supplier:
Avantor
CAS Number:
Specifications
Pk:
1 gal US
Seq:
8080
Specifications
About this item
Product Details
:
Product Details