Biological safety cabinet, Class II, type A2
Supplier: HAIER MEDICAL LABORATORY PRO
Certificates
About this item
The Haier biomedical biological safety cabinets are designed to protect the operator, laboratory environment and samples from being exposed to the infective aerosol produced from samples with bacteria strains, diagnostic materials and other infective substances. The units provide the operator with comfortable and safe working conditions. They are widely used in medical health, disease prevention, food safety, biological pharmacy and environment monitoring.
- Reverse centrifugal fan
- Double side air inflow design decreases running noise
- Air velocity auto compensation function guarantees stable wind speed
- Provide uniform airflow by adjusting voltage of fan
- 304 stainless steel operation platform and internal wall
- Ultra low penetration air filtration system-tested to a typical efficiency of 99,9995% for 0,12 µm particles
- Provide FED STD 209E Class 1 (or ISO 14644.1 Class 3) clean air to work surface in a stable vertical laminar flow to protect samples
- The exhaust ULPA filter traps biohazard particles acquired from the work surface before air is exhausted to the room, offering personnel and environmental protection
- High efficiency blower system
Power: 1300 VA
Certifications: EN 12469
Ordering information: Unit supplied with support stand.
Specifications
- Catalog No:
- HAIEHR40-IIA2
- HAIEHR30-IIA2
- Filters:
- Main filter typical efficiency: ULPA, 99,9995% at 0,12 μm
Exhaust filter typical effiency: HEPA, 99,995% at 0,3 μm - Main filter typical efficiency: ULPA, 99,9995% at 0,12 μm
Exhaust filter typical effiency: HEPA, 99,995% at 0,3 μm
- Average air velocity:
- Downflow velocity: 0,28 m/s
Inflow velocity: 0,55 m/s - Downflow velocity: 0,31 m/s
Inflow velocity: 0,55 m/s
- Lighting system:
- ≥1200 lux
- ≥1100 lux
- Ext. W×D×H:
- 1360×780×2200 mm
- 1100×790×2200 mm
- Int. W×D×H:
- 1167×610×680 mm
- 900×610×680 mm
Specifications
Frequently Bought Together