Order Entry
India
Orders LinkContactUsLinkComponent
Masterflex® Single-Use Sampling Bottle Cap Assembly, Avantor®
Masterflex® Single-Use Sampling Bottle Cap Assembly, Avantor®
Masterflex® Single-Use Sterile Sampling Bottle Assembly, PETG Bottle, 2 L, 53 mm, White, HDPE, 1/8" ID x 1/4" OD
Catalog # MFLX95644-32
Masterflex® Single-Use Sampling Bottle Cap Assembly, Avantor®
Catalog # MFLX95644-32
Masterflex® Single-Use Sampling Bottle Cap Assembly, Avantor®
Masterflex® Single-Use Sampling Bottle Cap Assembly, Avantor®
Masterflex® Single-Use Sterile Sampling Bottle Assembly, PETG Bottle, 2 L, 53 mm, White, HDPE, 1/8" ID x 1/4" OD
Price
Rs.8,500.00
Log in for availability
1 * 1 items
Product Details & Documents
Safely use your own bottles to transfer and store fluids in your work area.
  • Create a closed system to collect fluids from your application
  • Create a system with ventilation or to prevent spills
Single use cap assembly for connecting your bottles to your application and collect, transfer and store fluids. Cap assemblies come with Puri-Flex™ biopharma tube/linear assembly with 1/8" ID x 1/4" OD tubing and HDPE cap to fit a variety of bottles. Purchase bottle separately.
Delivery information: Includes cap and tubing.
Specifications
  • Cap Size:
    53 mm
  • Tubing int.Ø:
    1/8" ID x 1/4" OD
  • Colour:
    White
  • Description:
    Masterflex® Single-Use Sterile Sampling Bottle Assembly, PETG Bottle, 2 L
  • Material:
    HDPE